missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 9001 Full-Finger Impact Gloves with AVC™ Padding

Catalog No. 19820233
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820233 2X-Large
19820219 Small
19820229 Medium
19820230 Large
19820232 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19820233 Supplier Ergodyne™ Supplier No. 17776

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AVC™ padding in the palm and other key areas helps to reduce the effect of shocks and impacts. Full finger design enhances protection without sacrificing dexterity.

  • Segmented AVC™ padding reduces shock and impact
  • Gripping palm provides a secure hold on tools and equipment
  • Full-finger design enhances protection while maintaining dexterity
  • Hex stretch knit and breathable spandex construction
  • Neoprene knuckle pad with a reinforced thumb saddle
  • Low-profile molded hook & loop closure
  • Gloves feature ID space to write user's name

Specifications

Cuff Material Neoprene
Color Black
Hand Specific
Material Spandex
Product Type Anti-impact Gloves
Size 2X-Large
For Use With (Application) Oil/Gas Drilling, Mining, and Heavy Construction
Glove Cuff Style Hook and Loop
Series ProFlex™ 9001