missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 900 Half-Finger Impact Gloves

Catalog No. 19820198
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820198 Small
19820199 Medium
19820200 Large
19820201 X-Large
19820202 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19820198 Supplier Ergodyne™ Supplier No. 17692

AVC™ padding in the palm and other key areas helps to reduce shock and impact. Coating on the palm provides enhanced gripping capabilities.

  • Segmented AVC™ palm padding reduces shock and palm impact
  • Gripping palm provides secure hold on tools and equipment
  • Half-finger design for dexterity and tactile feedback
  • Hex stretch knit and breathable spandex construction
  • Neoprene knuckle pad with a reinforced thumb saddle
  • Low-profile molded hook & loop closure
  • Gloves include ID space to write user's names
  • Finger webbing loops for easy on/off

Specifications

Color Black
Hand Specific
Material Spandex
Product Type Anti-impact Gloves
Size Small
For Use With (Application) Construction, Manufacturing, Hand and Power Tools
Glove Cuff Style Hook and Loop
Series ProFlex™ 900