missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 500 Elbow Support

Catalog No. 2961186
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
2961186 Large
2961185 Medium
2961184 Small
19035285F X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 2961186 Supplier Ergodyne™ Supplier No. ERG16004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for jobs requiring repetitive arm motions

  • Wide base of support directs pressure away from tendons for improved performance with maximum comfort
  • Fully adjustable for a secure, exact fit
  • Wear over clothing or next to skin
  • Machine washable
  • Black

Specifications

Color Black
Product Type Elbow Support
Design Contoured
Size Large
For Use With (Application) Jobs requiring repetitive arm motions
Hand Ambidextrous