missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 380 Standard Kneeling Pad

Catalog No. 19820283
Change view
Click to view available options
Color: Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820283 Color: Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820283 Supplier Ergodyne™ Supplier No. 18381

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProFlex™ 380 Standard Kneeling Pad constructed of nitrile rubber (NBR) closed cell foam is designed to keep its shape over time.

  • Made with high density, thick NBR foam rubber that will not compress over time, protecting knees against hard surfaces
  • Water resistant and petroleum resistant construction
  • Kneeling pad is lightweight and has a built-in handle making it easy to carry and transport
  • Use to support and alleviate pressure from other areas like elbows, shoulders, and hips

Specifications

Color Orange
Size 14 x 21 x 1 in. (36 x 53 x 2.5 cm)
For Use With (Application) Plumbing, Mechanics, Utilites, Electricians, Landscaping, Carpentry
Material Nitrile Butadiene Rubber