missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 367 Round Cap Lightweight Gel Knee Pads

Catalog No. 19820291
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19820291 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820291 Supplier Ergodyne™ Supplier No. 18467

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProFlex™ 367 Round Cap Lightweight Gel Knee Pads provide lightweight gel cushioning and mobility.

  • Round cap for highest degree of kneeling mobility
  • Non-slip rubber cap grips kneeling surface
  • Lightweight gel and EVA foam for mobile comfort
  • Breathable perforated neoprene bottom strap
  • Top elastic strap cinches tight with plastic clip

Specifications

Color Black
Design Round Cap
For Use With (Application) Construction, Maintenance, Mechanics