missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 365 Mini Kneeling Pad

Catalog No. 19820281
Change view
Click to view available options
Includes:
Carabiner
Grabber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19820281 Grabber
19820290 Carabiner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19820281 Supplier Ergodyne™ Supplier No. 18465

ProFlex™ 365 Mini Kneeling Pad constructed of nitrile rubber (NBR) closed cell foam is designed to keep its shape over time.

  • Small and lightweight for optimal portability and storage
  • Built-in handle and carabiner for hanging storage or easy attachment
  • 1 in. (25 mm) NBR foam padding is nonconductive and keeps its shape over time
  • Water resistant and petroleum resistant construction
  • Use to support and alleviate pressure from other areas like elbows, shoulders, and hips

Specifications

Color Black
Dimensions (L x W) 4 x 6 in. (10 x 15 cm)
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Includes Grabber
Material Nitrile Butadiene Rubber