missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 357 Short Cap Lightweight Gel Knee Pads

Catalog No. 19820289
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820289 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820289 Supplier Ergodyne™ Supplier No. 18457

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProFlex™ 357 Short Cap Lightweight Gel Knee Pads provide lightweight gel cushioning and mobility in a short cap design.

  • Short cap design for mobility when kneeling
  • Non-slip rubber cap grips kneeling surface
  • Lightweight gel and EVA foam for mobile comfort
  • Bottom webbing and top elastic straps
  • Plastic clips cinch tight and keep straps secure

Specifications

Color Black
Design Short Cap
For Use With (Application) Construction, Maintenance, Mechanics