missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 347 Long Cap Lightweight Gel Knee Pads

Catalog No. 19820287
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820287 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820287 Supplier Ergodyne™ Supplier No. 18447

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProFlex™ 347 Long Cap Lightweight Gel Knee Pads provide lightweight gel cushioning, mobility, full knee coverage, and stability.

  • Long cap design provides protection and stability
  • Non-slip rubber cap grips kneeling surface
  • Lightweight gel and EVA foam for mobile comfort
  • Bottom webbing and top elastic straps
  • Plastic clips cinch tight and keep straps secure

Specifications

Color Black
Design Long Cap
For Use With (Application) Construction, Maintenance, Mechanics