missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 345 Long Cap Injected Gel Knee Pads

Catalog No. 19820286
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820286 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820286 Supplier Ergodyne™ Supplier No. 18445

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProFlex™ 345 Long Cap Injected Gel Knee Pads have perforated bottom straps and top elastic straps to keep the knee pad in position.

  • Long cap design provides protection and stability
  • Non-slip rubber cap grips kneeling surface
  • Injected gel and PU foam for all-day comfort
  • Breathable perforated neoprene bottom strap
  • Top elastic strap cinches tight with plastic clip

Specifications

Color Black
Design Long Cap
For Use With (Application) Construction, Maintenance, Flooring, Mechanics