missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 342 Extra Long Cap Injected Gel Knee Pads

Catalog No. 19820284
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820284 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820284 Supplier Ergodyne™ Supplier No. 18442

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProFlex™ 342 Extra Long Cap Injected Gel Knee Pads have extra long caps for maximum coverage and stability.

  • Extra long cap for maximum coverage and stability
  • Non-slip rubber cap grips kneeling surface
  • Injected gel and PU foam for all-day comfort
  • Breathable perforated neoprene top strap
  • Bottom elastic strap secures knee pad around calf
  • Extended shin protection optimizes stability

Specifications

Color Black
Design Extra Long Cap
For Use With (Application) Construction, Maintenance, Flooring, Mechanics