missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 1051 Mesh Back Support Brace

Catalog No. 19820392
Change view
Click to view available options
Size:
2X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820392 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820392 Supplier Ergodyne™ Supplier No. 20187

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProFlex™ 1051 Mesh Back Support Brace is constructed of cool mesh for lower back support.

  • Innovative Power Panel Positioners™
  • Breathable, mesh outer shell
  • Contoured ventilated lumbar pad
  • Two-stage closure

Specifications

Color Black
Product Type Innovative Power Panel Positioners™
Size 2X-Large
For Use With (Application) Material Handling, Construction, Warehousing/ Distribution, Maintenance, Landscaping