missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 100HV Economy Spandex Hi-Vis Back Support Brace

Catalog No. p-7163131
Change view
Click to view available options
Size:
2X-Large
3X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19819470 2X-Large
19819471 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19819470 Supplier Ergodyne™ Supplier No. 11886
Only null left

ProFlex™ 100HV Economy Spandex Hi-Vis Back Support designed to remind workers of proper lifting techniques while moving stress away from the spine for back pain relief.

  • High visibility
  • Polypropylene stays for long-lasting integrity
  • Adjustable and removable 0.5 to 1 in. (1 to 2.5 cm) shoulder straps keep the belt on standby until the next lift
  • Stretchable bias binding with zig-zag stitching
  • Rubber track webbing prevents riding up
  • Machine wash cold, air dry

Specifications

Color Orange
Size 2X-Large
For Use With (Application) Material Handling, Construction, Maintenance