missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 100 Economy Spandex Back Support Brace

Catalog No. 19819458
Change view
Click to view available options
Size:
X-Small
3X-Large
4X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19819458 X-Small
19819460 3X-Large
19819461 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19819458 Supplier Ergodyne™ Supplier No. 11381

ProFlex™ 100 Economy Spandex Back Support is designed to remind workers of proper lifting techniques while also moving stress away from the spine for back pain relief.

  • Stretchable spandex body
  • Polypropylene stays for long-lasting integrity
  • Adjustable, detachable suspenders keep belt in place
  • Rubber track webbing prevents riding up
  • Machine wash cold, air dry

Specifications

Color Black
Size X-Small
For Use With (Application) Material Handling, Construction, Maintenance
Material 280D Spandex
Size (English) Waist 25 to 58 in.
Size (Metric) Waist 64 to 147 cm