missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Proflavine hydrochloride

Catalog No. AC465860250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465860250 25 g
AC465860050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465860250 Supplier Thermo Scientific Chemicals Supplier No. 465860250
Only null left

CAS: 952-23-8 | C26H26Cl2N6O | 509.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 952-23-8
Molecular Formula C26H26Cl2N6O
Molecular Weight (g/mol) 509.44
MDL Number MFCD00066867
InChI Key APOVFGFHTIKDOX-UHFFFAOYSA-N
IUPAC Name dihydrogen bis(acridine-3,6-diamine) hydrate dichloride
SMILES [H+].[H+].O.[Cl-].[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1

Specifications

Alpha Vector PROFLAVINEHYDROCHLORIDE
Melting Point 270°C
Color Brown-Red to Orange
Loss on Drying ≤4 % (105°C, 1 hr)
Infrared Spectrum Conforms
Assay Percent Range 13.6 to 15.0 % (CL) (Titration Argentometric)
Packaging glass bottle
Quantity 25 g
Solubility Information (1 mg/ml aq. soln.) Clear solution
Formula Weight 245.71
Physical Form Powder
Chemical Name or Material Proflavine hydrochloride
Show More Show Less
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only