missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Productainer™ BioProcess Container (BPC), 50L with 4 Top Ports, 3D Single-Use BPC
GREENER_CHOICE

Catalog No. SH3065101
Change view
Click to view available options
Quantity:
50 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3065101 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3065101 Supplier Thermo Scientific™ Supplier No. SH30651.01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Using these BPC bags with 4 top ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

Using Thermo Scientific™ Productainer™ BPC bag with 4 top ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function Productainers can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - Collection and storage of bulk product from bioreactors

  • Storage - Sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation systems

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Description 50 L dip tube: 38 cm (15 in.)
Film Description CX5-14
Port Detail Top Ports: 2 x 0.25 in. 2 x 0.38 in.
For Use With (Equipment) SV50076.02
Material Polyethylene
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity 50 L
Sterilization Method Gamma Irradiated
Tubing Line 1 Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.) with dip tube length dependent on size
Tubing Line 2 Line 2. Insert luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
Tubing Line 3 Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
Tubing Line 4 Line 4. MPC body, polycarbonate Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.) dip tube length: 30.5 cm (12 in.)
Product Line Productainer
Show More Show Less