missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Productainer™ BioProcess Container (BPC), 50 L with 4 Top and 1 Bottom Ports, 3D Single-Use
GREENER_CHOICE

The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 50 L 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 4 top ports and 1 bottom port. The 3D square tube design with bottom port dispense is for use with conical, bottom-drain drums.

Supplier:  Thermo Scientific™ SH30652.01

Catalog No. SH3065201


Only null left

Description

Description

Features:

  • Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
  • All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
  • All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.

Applications of the Thermo Scientific 3D BPC include:

  • Hydration and sterilization of process buffers, liquids, and culture media.
  • Chromatography feed and fraction collection.
  • Storage and transport of bulk drug product and bulk drug precursors.
  • Harvest from and feed into bioreactors and fermentors.
  • Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology. Choose from the industry’s largest component library – with over 20,000 unique components that include:

  • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
  • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
  • Filters—used for bioburden reduction; sterilization grade filters and vent filters.
  • Tubing—thermoplastic elastomers, platinum-cured silicone.
  • Sample ports—septum-style and needle-free ports.
  • Clamps—sliders, pinch, and crimp bands.
Specifications

Specifications

50 L
Bottom Port: 1 x 0.38 in., Top Ports: 1 x 0.38 in. 2 x 0.25 in.
Packages placed in strong cardboard cartons with product and lot identification
50 L
Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 121.9 cm (48 in.) dip tube ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) with dip tube length: 30.5 cm (12 in.) Top Port
Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port
Line 4. Plugged, no tubing
CX5-14
SV50517.04
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Gamma Irradiated
Line 2. Insert luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port
Line 5. MPC body, polycarbonate Tubing: silicone, length: 121.9 cm (48 in.) ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) Bottom port
Productainer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions