missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Productainer™ BioProcess Container (BPC), 200L with 3 Ports, 2D BPC

Description
The Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 200 L 2D pillow design is composed of CX5-14 single web, multi-layer plastic film and includes 3 panel ports.
Features:
- Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
- All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.
Applications of the Thermo Scientific Labtainer BPC include:
- Harvest from, feed into, and collect samples from bioreactors and fermentors.
- Buffer, culture media, and process liquid preparation and storage.
- Chromatography feed and fraction collection.
- Harvest, storage and transport of bulk drug product and bulk drug precursors.
Customize standard BPC systems for optimized single-use technology
Choose from the industry's largest component library – with over 20,000 unique components that include:
- Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
- Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
- Filters—used for bioburden reduction; sterilization grade filters and vent filters.
- Tubing—thermoplastic elastomers, platinum-cured silicone.
- Sample ports—septum-style and needle-free ports.
- Clamps—sliders, pinch, and crimp bands.
Specifications
Specifications
| Green Features | ISCC PLUS |
| Capacity (Metric) | 200 L |
| Film Description | CX5-14 |
| Port Detail | Panel Ports: 2 x 0.38 in. 1 x 0.25 in. |
| For Use With (Equipment) | HyPerforma Flat-bottom LLDPE Drum - SV50076.04 |
| Dimensions | Chamber: 94 cm x 142 cm (37.5 in. x 56 in.) |
| Material | Polyethylene |
| Outer Packaging | Packages placed in strong cardboard cartons with product and lot identification |
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
| Quantity | 1 |
| Show More |