missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Productainer™ 3D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, MPXs
Description
Thermo Scientific™ Productainer 3D BioProcess Containers (BPCs) are used to package, store, and transport media, buffers, water for injection (WFI), process liquids, and biological products. Productainer BPCs are constructed with Aegis 5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainer BPCs are suitable for a wide array of bioprocess applications. For optimal fit, form, and function, Productainer BPCs can be customized using the industry's largest component library.
Highlights:
- Film optimized for flexibility and durability—superior stress resistance, puncture resistance, and physical barrier properties to ensure stability of contents
- Gamma-irradiation sterilized
- Squared tube design with polyethylene ports located on the top of the BPC for easy access
- Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
- Customizable via the industry's largest library of qualified components
Recommended for:
- Harvest – collection and storage of bulk product from bioreactors
- Storage – sterile storage of media, buffers, and process intermediates
- Separation – feeding and receiving of sterile liquids from separation systems
- Purification – buffer feeding to purification system, fraction collection and storage
- Filling – sterile reservoir for filling systems
- Transport and shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
Specifications
| Capacity (Metric) | 500 L |
| Quantity | 500 L |
| Tubing Style | Platinum-cured Silicone |