missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Productainer™ 3D BioProcess Container (BPC), Aegis™ 5-14 film, five ports, Luer locks + MPCs

Catalog No. SH3096904
Change view
Click to view available options
Capacity (Metric):
500 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3096904 500 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3096904 Supplier Thermo Scientific™ Supplier No. SH30969.04

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Productainer 3D BioProcess Containers (BPCs) are used to package, store, and transport media, buffers, water for injection (WFI), process liquids, and biological products.

Thermo Scientific™ Productainer 3D BioProcess Containers (BPCs) are used to package, store, and transport media, buffers, water for injection (WFI), process liquids, and biological products. Productainer BPCs are constructed with Aegis 5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainer BPCs are suitable for a wide array of bioprocess applications. For optimal fit, form, and function, Productainer BPCs can be customized using the industry's largest component library.

Highlights:

  • Film optimized for flexibility and durability—superior stress resistance, puncture resistance, and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest – collection and storage of bulk product from bioreactors
  • Storage – sterile storage of media, buffers, and process intermediates
  • Separation – feeding and receiving of sterile liquids from separation systems
  • Purification – buffer feeding to purification system, fraction collection and storage
  • Filling – sterile reservoir for filling systems
  • Transport and shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Capacity (Metric) 500 L
Quantity 500 L
Tubing Style C-Flex