Learn More
Thermo Scientific™ Productainer™ 3D BioProcess Container, BioTitan retainers, two ports, MPCs

Use Thermo Scientific Productainer BPC bags with BioTitan retainers to package, store, and transport media, buffers, water for injection (WFI), process liquids, and biological products.
$494.36 - $699.25
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
SH3129701
![]() |
Thermo Scientific™
SH31297.01 |
50 L |
Each for $494.36
|
|
|||||
|
SH3129702
|
Thermo Scientific™
SH31297.02 |
100 L |
Each for $699.25
|
|
|||||
SH3129703
![]() |
Thermo Scientific™
SH31297.03 |
200 L |
Each for $578.50
|
|
|||||
Description
Productainer BPCs are constructed with our Thermo Scientific Aegis5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.
Features include:
- Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
- Gamma-irradiation sterilized
- Squared tube design with polyethylene ports located on the top of the BPC for easy access
- Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
- Customizable via the industry's largest library of qualified components
- Now manufactured with the BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
Recommended for:
- Harvest—collection and storage of bulk product from bioreactors
- Storage—sterile storage of media, buffers and process intermediates
- Separation—feeding and receiving of sterile liquids from separation systems
- Purification—buffer feeding to purification system, fraction collection and storage
- Filling—sterile reservoir for filling systems
- Transport and Shipping—bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
| TPE |