Learn More
Agilent BioTek Product Qualification Package
Catalog No. BT1940556
Change view
Click to view available options
For Use With (Equipment):
Cytation C10
Catalog No. For Use With (Equipment)
BT1940556 Cytation C10
1 options
Note: Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
Catalog No. BT1940556 Supplier Agilent BioTek Supplier No. 1940556
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Note: Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
Product qualification package for use with Cytation C10
  • Product qualification package
  • For Cytation C10
Specifications
For Use With (Equipment) Cytation C10