missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synbiosis™ ProDilute Gravimetric Diluter
Description
ProDilute dilutes the sample by adjusting the delivered volume to the weight of the sample. Automatic distribution of the broth or media guarantees precision of the dilution.
- Easy loading of bags or samples
- Rapid dispensing- less than 10 seconds to dispense 225 mL
- Compact and light at 8 kg
- Minimal and flexible bench space
- Ambidextrous controller
- Unique weighing reliability
- Built-in bubble level and adjustable legs
- Weighing independent of the positioning of the sample in its holder
- Finger-stop design to prevent contamination of distribution nozzle during installation
Specifications
Specifications
| Description | Gravimetric Diluter |
| Dimensions (L x W x H) | 27.5 x 25 x 40 cm |
| Voltage | 230 V |
| Wattage | 120 W |
| Frequency | 50 Hz |