missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProClean™ Coverall (all)

Catalog No. 19088360
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
5 to 5.7 ft.
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
6.3 to 7 ft.
Height (Metric):
152.4 to 173.73 cm
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
192.02 to 213.3 cm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
19088360 4X-Large 6.3 to 7 ft. 192.02 to 213.3 cm
19088354 Small 5 to 5.7 ft. 152.4 to 173.73 cm
19088359 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
19088355 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
19088358 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
19088357 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
19088356 Large 5.8 to 6 ft. 176.78 to 182.88 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19088360 Supplier DuPont™ Supplier No. PC143SWH4X00250B

Unique breathable composite material provides particulate and light splash protection for critical environments. Coveralls feature a zipper front, serged seams, and elastic wrists, ankles, and waist.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Coverall has elastic openings for tighter fit at wrist and ankle.
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing.
  • Enclosed elastic at the back of the garment for a better fit
  • Bulk packaged in double transparent poly liners.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • D13477891,D13477875,D13477852,D13477915,D13477791,D13477836

Specifications

Product Type Cleanroom Coveralls
Size 4X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Back Style Elastic
Size (English) Chest 52 to 58 in.
Size (English) Inseam 33.5 in.
Size (Metric) Chest 132.08 to 147.32 cm
Size (Metric) Inseam 85.09 cm
Sleeve Style Set
Thickness 5.4 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
Length (English) Sleeve 41.5 in.
Length (Metric) Sleeve 105.41 cm
Height (English) 6.3 to 7 ft.
Height (Metric) 192.02 to 213.3 cm
Packaging Bulk
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less