missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Procion Blue H-5R

Catalog No. AAA1271922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1271922 100 g
AAA1271914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1271922 Supplier Thermo Scientific Chemicals Supplier No. A1271922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12236-84-9

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12236-84-9
MDL Number MFCD00070851
Synonym C.I. 181575; Reactive Blue 13

Specifications

Quantity 100 g
Chemical Name or Material Procion Blue H-5R
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only