missing translation for 'onlineSavingsMsg'
Learn More

Prochlorperazine Dimaleate 98.0+%, TCI America™

Catalog No. P23685G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23685G Supplier TCI America Supplier No. P23685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prochlorperazine dimaleate for use as a laboratory research reagent.

Specifications

CAS 84-02-6, 58-38-8
Melting Point 194°C
Color White-Yellow
Molecular Formula C28H32ClN3O8S
Quantity 5 g
MDL Number MFCD00069330
Synonym prochlorperazine dimaleate, prochlorperazine maleate, unii-i1t8o1jtl6, buccastem, emetiral, pasotomin, prochlorperazine dimaleate salt, nibromin a, meterazin maleate, stemetil dimaleate
InChI Key DSKIOWHQLUWFLG-SPIKMXEPSA-N
SMILES CN1CCN(CC1)CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)Cl.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;2-chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine
Molecular Weight (g/mol) 606.087
PubChem CID 5281032
ChEBI CHEBI:8436
Formula Weight 606.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Prochlorperazine Dimaleate
Show More Show Less
RTECSNumber SO3150000
TSCA No