Learn More
Miele Inc ProCare Lab 30 C - 10 L
Catalog No. 501713094
Encompass
ProCare Lab 30 C - 10 LNeutralizing agent, acidic, 10 l for optimum neutralization with citric acid based neutralizing agent.Free from surfactants, phosphorus compounds, and oxidation agentsSpecially developed for Miele ProfessionalSuitable for all types of materialsTo be used as neutralizing agent or acid pre-cleanerpH value 2.6-3
Catalog No. 501713094 Supplier Miele Inc Supplier No. 10266680
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ProCare Lab 30 C - 10 LNeutralizing agent, acidic, 10 l for optimum neutralization with citric acid based neutralizing agent.Free from surfactants, phosphorus compounds, and oxidation agentsSpecially developed for Miele ProfessionalSuitable for all types of materialsTo be used as neutralizing agent or acid pre-cleanerpH value 2.6-3