missing translation for 'onlineSavingsMsg'
Learn More

Procarbazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135598
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2550100MG 100 mg
P2550500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2550100MG Supplier TCI America Supplier No. P2550100MG
Only null left

Chemical Identifiers

CAS 366-70-1
Molecular Formula C12H20ClN3O
Molecular Weight (g/mol) 257.762
MDL Number MFCD00072082
InChI Key DERJYEZSLHIUKF-UHFFFAOYSA-N
Synonym procarbazine hydrochloride, procarbazine hcl, matulane, nathulane, natunalar, mih hydrochloride, pcb hydrochloride, procarbazine.hcl, ibenzmethyzine hydrochloride, natulan hydrochloride
PubChem CID 9703
ChEBI CHEBI:71428
IUPAC Name 4-[(2-methylhydrazinyl)methyl]-N-propan-2-ylbenzamide;hydrochloride
SMILES CC(C)NC(=O)C1=CC=C(C=C1)CNNC.Cl

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 100 mg
Formula Weight 257.76
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Procarbazine Hydrochloride
EINECSNumber (3)-2979
RTECSNumber XS4725000
TSCA No