missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procaine 98.0+%, TCI America™
$90.48 - $269.52
Chemical Identifiers
| CAS | 59-46-1 |
|---|---|
| Molecular Formula | C13H20N2O2 |
| Molecular Weight (g/mol) | 236.315 |
| MDL Number | MFCD00007893 |
| InChI Key | MFDFERRIHVXMIY-UHFFFAOYSA-N |
| Synonym | procaine, vitamin h3, novocaine, spinocaine, duracaine, procain, nissocaine, novocain, 2-diethylamino ethyl 4-aminobenzoate, procaina |
| PubChem CID | 4914 |
| ChEBI | CHEBI:8430 |
| IUPAC Name | 2-(diethylamino)ethyl 4-aminobenzoate |
| SMILES | CCN(CC)CCOC(=O)C1=CC=C(C=C1)N |
Chemical Identifiers
| 59-46-1 | |
| 236.315 | |
| MFDFERRIHVXMIY-UHFFFAOYSA-N | |
| 4914 | |
| 2-(diethylamino)ethyl 4-aminobenzoate |
| C13H20N2O2 | |
| MFCD00007893 | |
| procaine, vitamin h3, novocaine, spinocaine, duracaine, procain, nissocaine, novocain, 2-diethylamino ethyl 4-aminobenzoate, procaina | |
| CHEBI:8430 | |
| CCN(CC)CCOC(=O)C1=CC=C(C=C1)N |
Specifications
| 59-46-1 | |
| White | |
| MFCD00007893 | |
| procaine, vitamin h3, novocaine, spinocaine, duracaine, procain, nissocaine, novocain, 2-diethylamino ethyl 4-aminobenzoate, procaina | |
| CCN(CC)CCOC(=O)C1=CC=C(C=C1)N | |
| 236.315 | |
| CHEBI:8430 | |
| ≥98.0% (HPLC,T) | |
| Procaine |
| 62°C | |
| C13H20N2O2 | |
| 5 g | |
| MFDFERRIHVXMIY-UHFFFAOYSA-N | |
| 2-(diethylamino)ethyl 4-aminobenzoate | |
| 4914 | |
| 236.32 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0162
RTECSNumber : DG2100000
TSCA : No