missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procaine Hydrochloride 98.0+%, TCI America™
$42.87 - $113.71
Chemical Identifiers
| CAS | 51-05-8 |
|---|---|
| Molecular Formula | C13H21ClN2O2 |
| Molecular Weight (g/mol) | 272.77 |
| MDL Number | MFCD00013000 |
| InChI Key | HCBIBCJNVBAKAB-UHFFFAOYSA-N |
| Synonym | procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor |
| PubChem CID | 5795 |
| ChEBI | CHEBI:8431 |
| IUPAC Name | hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride |
| SMILES | [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1 |
Chemical Identifiers
| 51-05-8 | |
| 272.77 | |
| HCBIBCJNVBAKAB-UHFFFAOYSA-N | |
| 5795 | |
| hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride |
| C13H21ClN2O2 | |
| MFCD00013000 | |
| procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor | |
| CHEBI:8431 | |
| [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1 |
Specifications
| 51-05-8 | |
| White | |
| MFCD00013000 | |
| 2811 | |
| HCBIBCJNVBAKAB-UHFFFAOYSA-N | |
| hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride | |
| 5795 | |
| 272.77 | |
| Crystalline Powder |
| 156°C | |
| C13H21ClN2O2 | |
| 25 g | |
| procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor | |
| [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1 | |
| 272.77 | |
| CHEBI:8431 | |
| ≥98.0% (HPLC,T) | |
| Procaine Hydrochloride |
Safety and Handling
RTECSNumber : DG2275000
TSCA : Yes