missing translation for 'onlineSavingsMsg'
Learn More

Procaine Hydrochloride 98.0+%, TCI America™

Catalog No. A116325G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A116325G 25 g
A1163100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A116325G Supplier TCI America Supplier No. A116325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51-05-8
Molecular Formula C13H21ClN2O2
Molecular Weight (g/mol) 272.77
MDL Number MFCD00013000
InChI Key HCBIBCJNVBAKAB-UHFFFAOYSA-N
Synonym procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor
PubChem CID 5795
ChEBI CHEBI:8431
IUPAC Name hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride
SMILES [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 156°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 272.77
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Procaine Hydrochloride
RTECSNumber DG2275000
TSCA Yes