missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Procaine hydrochloride, 99%

Catalog No. AC207315000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207315000 500 g Plastic bottle
AC207311000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC207315000 Supplier Thermo Scientific Chemicals Supplier No. 207315000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51-05-8 | C13H21ClN2O2 | 272.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-05-8
Molecular Formula C13H21ClN2O2
Molecular Weight (g/mol) 272.77
MDL Number MFCD00013000
InChI Key HCBIBCJNVBAKAB-UHFFFAOYSA-N
Synonym procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor
PubChem CID 5795
ChEBI CHEBI:8431
IUPAC Name 2-(diethylamino)ethyl 4-aminobenzoate;hydrochloride
SMILES [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 154.0°C to 158.0°C
Color Colorless to White
pH 5 to 6.5 (2% w/v aq. soln.)
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NC6H4CO2CH2CH2N(C2H5)2·HCl
Quantity 500 g
Beilstein 14, 424
Merck Index 15, 7875
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform, practically insoluble in ether
Formula Weight 272.77
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Procaine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-077-2

RUO – Research Use Only