missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Probe Set Diluent QT

Catalog No. QVT0510
Change view
Click to view available options
Quantity:
12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QVT0510 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QVT0510 Supplier Invitrogen™ Supplier No. QVT0510
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Probe Set Diluent QT, 12 mL is an aqueous solution containing formamide, detergent, and blocker.

Probe Set Diluent QT, 12 mL is an aqueous solution containing formamide, detergent, and blocker. It is an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.

Specifications

Content And Storage Store at 2°C to 8°C
Format Kit
Product Type Probe Set Diluent
Quantity 12 mL

For Research Use Only. Not for use in diagnostic procedures.