missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Proadifen hydrochloride, 95%

Catalog No. AC457880010
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457880010 1g
AC457880050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457880010 Supplier Thermo Scientific™ Supplier No. 457880010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 62-68-0
Molecular Formula C23H31NO2·HCl
Molecular Weight (g/mol) 389.96
InChI Key FHIKZROVIDCMJA-UHFFFAOYSA-N
Synonym proadifen hydrochloride, proadifen hcl, propyladiphenin, skf-525a hydrochloride, skf 525a, SK&F 525-a, proadifen hydrochloride usan, skf 525-a, skf-525a, hydrochloride
PubChem CID 65341
IUPAC Name 2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
SMILES CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)OCCN(CC)CC.Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 1g
Solubility Information Solubility in water: soluble.
Formula Weight 389.96
Percent Purity 95%
Chemical Name or Material Proadifen hydrochloride

RUO – Research Use Only