missing translation for 'onlineSavingsMsg'
Learn More

Berkshire Pro-Wipe™ SuperSorb Dispenser box

Catalog No. 19063112
Change view
Click to view available options
Length (English):
9 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19063112 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19063112 Supplier Berkshire Supplier No. PWSS0914D16

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pro-Wipe™ Supersorb is tough enough to be a rag replacement and clean enough to be used in a controlled manufacturing environment.

  • No binders or other chemical additives
  • Smooth and soft with high strength and durability
  • Super heavy absorbency
  • Low linting and abrasion resistant
  • Chemically compatible with common cleaning and disinfecting solutions
  • Absorbs water and oil based solutions
  • Autoclavable

Specifications

Product Type Medium-duty All Purpose Industrial Wiper
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 14 in.
Width (Metric) 36 cm
Material Non-woven Cellulose, Lyocell, Bicomponent
Autoclavable Autoclavable
Disposable Single-use
For Use With (Application) Cleaning of lab equipment, General cleaning of manufacturing equipment, Spill pick up, Surface prep, Dispenser box
Sterility Non-sterile