Learn More
Carson™ PRO Series MeasureLoupe Lighted Loupe Magnifier
PRO Series MeasureLoupe Lighted Loupe Magnifier is an 11.5X power LED and UV lighted loupe magnifier with a reticle scale and storage case.
Supplier: Carson™ CP45
Description
- Contains a high powered 11.5X magnification
- Ideal for inspecting close details for numerous hobbies and tasks
- Lens is made of a high grade optical glass that contains an anti-reflection coating for sharp and distortion-free viewing
- Removable reticle scale is measured in both inches and millimeters
- Ideal lighted magnifier for jewelers, stamp and coin collectors, rock and nature study, identifying patterns on fabrics, counting threads, printing, examining film slides, model building, electronics, inspecting circuit boards for soldering defects, identifying counterfeit currency and other crafts and tasks.
- Powered by 3 AAA batteries (not included)
- Comes included with a zippered protective pouch with a carabiner clip attachment
- Protective pouch will protect the glass lens from damage, dust and debris
Specifications
| Reticle Scale and Storage Case | |
| LED and UV | |
| 11.5x | |
| Magnifier |