Learn More
Invitrogen™ Pro-Q™ Emerald 300 Lipopolysaccharide Gel Stain Kit
Pro-Q™ Emerald 300 Lipopolysaccharide Gel Stain Kit
Supplier:  Invitrogen™ P20495
Encompass
Catalog No. P20495
Only null left
Description
Description
Molecular Probes' proprietary Pro-Q Emerald 300 and Pro-Q Emerald 488 Glycoprotein Stain Kits provide the most advanced technology available for detection of glycoproteins in gels and on blots. Gel staining is rapid and very sensitive. In less than three hours, it is possible to detect as little as 300 pg of glycoprotein per band, depending on the degree of glycosylation, making these stains at least 50-fold more sensitive than standard fuchsin staining.
Specifications
Specifications
In-Blot Detection, In-Gel Detection
Pro-Q Emerald 300
Lipopolysaccharide
Lipopolysaccharide Stain Kit
Fluorescence
1 kit
PRO-Q
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 6 Months
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.