Learn More
Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Staining Kits
The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards.
Supplier:  Invitrogen™ MPP33300
Encompass
 View more versions of this product
Catalog No. MPP33300
Only null left
Description
Description
The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards. Pro-Q Diamond Phosphoprotein Gel Stain is a fluorescent stain (excitation/emission maxima ∼555/580 nm) that can detect as little as 1–16 ng of phosphoprotein per band with a signal that is linear over three orders of magnitude. The PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.
The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards. Pro-Q Diamond Phosphoprotein Gel Stain is a fluorescent stain (excitation/emission maxima ∼555/580 nm) that can detect as little as 1–16 ng of phosphoprotein per band with a signal that is linear over three orders of magnitude. PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.
Features include:
Sensitive—detection level down to 1–16 ng of phosphoprotein per band
Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis
Built-in controls—molecular weight marker allows for approximation of protein size and serves as positive and negative control with unique mixture of phosphorylated and non-phosphorylated proteins
Linear signal—exhibits excellent linearity over three orders of magnitude, ideal for quantitation
Specifications
Chemical Identifiers
In-Gel Detection
Pro-Q Diamond
PRO-Q
Fluorescence
Proteins (Phosphoproteins)
Phosphoprotein Gel Staining Kit
Specifications
• Pro-Q™ Diamond Phosphoprotein Gel Stain, 1 L
•PeppermintStick™ Phosphoprotein Molecular Weight Standard, 40 μL
In-Gel Detection
Pro-Q Diamond
Proteins (Phosphoproteins)
Phosphoprotein Gel Staining Kit
Pro-Q™ Diamond Phosphoprotein Gel Staining Kit, 1 L
Fluorescence
1 L Stain, 40 μL Standard Kit
PRO-Q
Product Suggestions
Product Suggestions