Learn More
Pro-Lab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System
Designed to simplify the storage and retrieval of bacterial and fungal cultures. Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.
Supplier:  Pro-Lab Diagnostics™ PL.170C/Y
Encompass
 View more versions of this product
Catalog No. 22286157
Only null left
Description
Description
Designed to simplify the storage and retrieval of bacterial and fungal cultures. Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.
  • Incorporates treated beads and a special cryopreservative solution
  • Each vial contains 25 beads, providing repeated identical cultures from the original organism
  • Specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries
  • 80 Vials supplied in a freezer box with“see through”cover
  • Available in 5 cap/bead colors or mixed (16 vials each of blue, green, light blue, red, and yellow)
Order Info
System supplied as box of 80 vials
Specifications
Chemical Identifiers
Cryogenic Storage Vial
Designed to simplify the storage and retrieval of bacterial and fungal cultures
80 Pk.
-70°C
Specifications
Cryogenic Storage Vial
Designed to simplify the storage and retrieval of bacterial and fungal cultures
-70°C
Yellow
80 Pk.
Product Suggestions
Product Suggestions