missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System

Catalog No. 22286155
Encompass
Change view
Click to view available options
Closure Color:
Assorted
Blue
Green
Light Blue
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color
22286155 Assorted
22286152 Green
22286157 Yellow
22286153 Blue
22286156 Light Blue
22286154 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22286155 Supplier Pro-Lab Diagnostics™ Supplier No. PL.170C/M
Only null left

Designed to simplify the storage and retrieval of bacterial and fungal cultures. Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.

  • Incorporates treated beads and a special cryopreservative solution
  • Each vial contains 25 beads, providing repeated identical cultures from the original organism
  • Specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries
  • 80 Vials supplied in a freezer box with“see through”cover
  • Available in 5 cap/bead colors or mixed (16 vials each of blue, green, light blue, red, and yellow)

Order Info

System supplied as box of 80 vials

Specifications

Product Type Cryogenic Storage Vial
Closure Color Assorted
For Use With (Application) Designed to simplify the storage and retrieval of bacterial and fungal cultures
Quantity 80 Pk.
Storage Requirements -70°C