missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System
Description
Designed to simplify the storage and retrieval of bacterial and fungal cultures. Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.
- Incorporates treated beads and a special cryopreservative solution
- Each vial contains 25 beads, providing repeated identical cultures from the original organism
- Specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries
- 80 Vials supplied in a freezer box with“see through”cover
- Available in 5 cap/bead colors or mixed (16 vials each of blue, green, light blue, red, and yellow)
Order Info
System supplied as box of 80 vials
Specifications
Specifications
| Product Type | Cryogenic Storage Vial |
| Closure Color | Green |
| For Use With (Application) | Designed to simplify the storage and retrieval of bacterial and fungal cultures |
| Quantity | 80 Pk. |
| Storage Requirements | -70°C |