Learn More
SignalChem Biotech PNMT, Active 500ug
Catalog No. 502573724
Encompass
Full-length recombinant human PNMT was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502573724 Supplier SignalChem Biotech Supplier No. P331380G500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Full-length recombinant human PNMT was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.