missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Standard MSDS Binder - 1.5 in. Ring Dia. Binder with Inserts

Catalog No. 19103245
Change view
Click to view available options
Width (Metric):
311.15 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19103245 311.15 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19103245 Supplier Brady™ Supplier No. BR823A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Store your company's critical material safety data sheets (MSDS) information in a highly visible MSDS binder.

  • Binder helps keep material safety data sheets readily available for quick, easy access
  • Durable polyethylene cover protects material safety data sheets
  • Includes a 36 in. chain and a pair of sheet lifters
  • Binder is yellow with red writing

Specifications

Format Binder
Includes Binder; Chain; Inserts; Pair of Sheet Lifters
Diameter (English) Ring 1.5 in.
Width (Metric) 311.15 mm
Quantity Each
Depth (English) 2 in.
Material Polyethylene
Languages English
For Use With (Application) Hazard Communication