missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Shoe Cover Dispenser

Catalog No. 19124567
Change view
Click to view available options
D x W x H: 8 x 12 x 14 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19124567 D x W x H: 8 x 12 x 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19124567 Supplier Brady™ Supplier No. PD322E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Remind employees to wear foot protection by mounting shoe cover dispensers near all hazardous work areas. The Brady™ Shoe Cover Dispenser features SHOE COVERS printed on the front for easy use, and can hold up to 50 covers. Covers are dispensed through an opening in the front.

  • Front opening of the dispenser gives convenient access to the items inside
  • Wall or table mounted
  • Made of clear acrylic with “SHOE COVERS” printed on a blue background
  • Package includes dispenser only (shoe covers sold separately)

Specifications

Product Type PPE Dispenser
Color Clear
Depth (English) 8 in.
Width (English) 12 in.
Height (English) 14 in.
Holds Shoe Covers
Material Acrylic
Mounting Wall or Surface Mount
No. of Compartments 1
Product Style Hinged Lid
Description Legend: “SHOE COVERS”
Capacity 50 Shoe Covers
For Use With (Application) Shoe cover storage & dispensing
Show More Show Less