missing translation for 'onlineSavingsMsg'
Learn More

Brady™ OSHA 300 BINDER & RACK

Catalog No. 19103200
Change view
Click to view available options
Width (Metric):
270.51 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19103200 270.51 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19103200 Supplier Brady™ Supplier No. OR314E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Our comprehensive OSHA Record Keeping Center offers you immediate compliance solutions and is sold as a complete kit (OR305E) or components sold separately

OSHA 300 binder and rack

Specifications

Format Information Center
Includes Binder; Rack
Width (Metric) 270.51 mm
Quantity 1 Ea.
Languages English
For Use With (Application) Facility information