missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Large Ball Valve Lockout Device, 2 to 8 in.

Catalog No. 19103899
Change view
Click to view available options
Color:
Blue
Green
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19103899 Blue
19103900 Green
19103901 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19103899 Supplier Brady™ Supplier No. BS08ABU

Patented design and ultra-tough polypropylene material make these devices the best choice to assuredly lock out your ball valves.

  • Large Ball Valve Lockout Device locks out pipes 2 in. to 8 in. in Dia.
  • Ultra-tough polypropylene resists temperatures from 0° to 175°F (-18° to 79°C)
  • Equipping your employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs
  • Brady™ is a world leader in lockout/tagout products and services offering industry expertise and a broad range of durable, easy-to-use lockout devices

Specifications

Product Type Valve Lockout Device
Color Blue
Quantity 1 Ea.