Learn More
Epredia™ PrintMate™ Slotted Cassettes
Catalog No. B851090BE
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Tan
White
Yellow
Quantity:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
B851090BE Beige 4 x 250 cassettes (1000)/cs
B851090BL Blue 4 x 250 cassettes (1000)/cs
B851090GN Green 4 x 250 cassettes (1000)/cs
B851090GY Gray 4 x 250 cassettes (1000)/cs
B851090LA Lavender 4 x 250 cassettes (1000)/cs
B851090LG Light Green 4 x 250 cassettes (1000)/cs
B851090LI Lilac 4 x 250 cassettes (1000)/cs
B851090OR Orange 4 x 250 cassettes (1000)/cs
B851090TN Tan 4 x 250 cassettes (1000)/cs
B851090WH White 4 x 250 cassettes (1000)/cs
B851090YW Yellow 4 x 250 cassettes (1000)/cs
B851120BE Beige 10 x 75 cassettes (750)/cs
B851120BL Blue 10 x 75 cassettes (750)/cs
B851120GN Green 10 x 75 cassettes (750)/cs
B851120GY Gray 10 x 75 cassettes (750)/cs
B851120LA Lavender 10 x 75 cassettes (750)/cs
B851120LG Light Green 10 x 75 cassettes (750)/cs
B851120LI Lilac 10 x 75 cassettes (750)/cs
B851120OR Orange 10 x 75 cassettes (750)/cs
B851120TN Tan 10 x 75 cassettes (750)/cs
B851120WH White 10 x 75 cassettes (750)/cs
B851120YW Yellow 10 x 75 cassettes (750)/cs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. B851090BE Supplier Epredia™ Supplier No. B851090BE
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Designed for performance and user convenience, PrintMate Slotted Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
Specifications
Pore Style Slots
Quantity 4 x 250 cassettes (1000)/cs
Closure Included Separate lids included
Closure Type Reversible
PrintMate Compatible Yes
Color Beige
Product Type Slotted Cassette