missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ PrintMate™ Embedding Cassettes

Catalog No. b851010la
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Quantity:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
Closure Included:
No lids
Separate lids included
48 product options available for selection
Product selection table with 48 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity Closure Included
B851010LA Lavender 10 x 75 cassettes (750)/cs No lids
B851010BE Beige 10 x 75 cassettes (750)/cs No lids
B851010BL Blue 10 x 75 cassettes (750)/cs No lids
B851010GN Green 10 x 75 cassettes (750)/cs No lids
B851010GY Gray 10 x 75 cassettes (750)/cs No lids
B851010LG Light Green 10 x 75 cassettes (750)/cs No lids
B851010LI Lilac 10 x 75 cassettes (750)/cs No lids
B851010OR Orange 10 x 75 cassettes (750)/cs No lids
B851010PK Pink 10 x 75 cassettes (750)/cs No lids
B851010TN Tan 10 x 75 cassettes (750)/cs No lids
B851010WH White 10 x 75 cassettes (750)/cs No lids
B851010YW Yellow 10 x 75 cassettes (750)/cs No lids
B851040BE Beige 4 x 250 cassettes (1000)/cs No lids
B851040BL Blue 4 x 250 cassettes (1000)/cs No lids
B851040GN Green 4 x 250 cassettes (1000)/cs No lids
B851040GY Gray 4 x 250 cassettes (1000)/cs No lids
B851040LA Lavender 4 x 250 cassettes (1000)/cs No lids
B851040LG Light Green 4 x 250 cassettes (1000)/cs No lids
B851040LI Lilac 4 x 250 cassettes (1000)/cs No lids
B851040OR Orange 4 x 250 cassettes (1000)/cs No lids
B851040PK Pink 4 x 250 cassettes (1000)/cs No lids
B851040TN Tan 4 x 250 cassettes (1000)/cs No lids
B851040WH White 4 x 250 cassettes (1000)/cs No lids
B851040YW Yellow 4 x 250 cassettes (1000)/cs No lids
B851070BE Beige 4 x 250 cassettes (1000)/cs Separate lids included
B851070BL Blue 4 x 250 cassettes (1000)/cs Separate lids included
B851070GN Green 4 x 250 cassettes (1000)/cs Separate lids included
B851070GY Gray 4 x 250 cassettes (1000)/cs Separate lids included
B851070LA Lavender 4 x 250 cassettes (1000)/cs Separate lids included
B851070LG Light Green 4 x 250 cassettes (1000)/cs Separate lids included
B851070LI Lilac 4 x 250 cassettes (1000)/cs Separate lids included
B851070OR Orange 4 x 250 cassettes (1000)/cs Separate lids included
B851070PK Pink 4 x 250 cassettes (1000)/cs Separate lids included
B851070TN Tan 4 x 250 cassettes (1000)/cs Separate lids included
B851070WH White 4 x 250 cassettes (1000)/cs Separate lids included
B851070YW Yellow 4 x 250 cassettes (1000)/cs Separate lids included
B851100BE Beige 10 x 75 cassettes (750)/cs Separate lids included
B851100BL Blue 10 x 75 cassettes (750)/cs Separate lids included
B851100GN Green 10 x 75 cassettes (750)/cs Separate lids included
B851100GY Gray 10 x 75 cassettes (750)/cs Separate lids included
B851100LA Lavender 10 x 75 cassettes (750)/cs Separate lids included
B851100LG Light Green 10 x 75 cassettes (750)/cs Separate lids included
B851100LI Lilac 10 x 75 cassettes (750)/cs Separate lids included
B851100OR Orange 10 x 75 cassettes (750)/cs Separate lids included
B851100PK Pink 10 x 75 cassettes (750)/cs Separate lids included
B851100TN Tan 10 x 75 cassettes (750)/cs Separate lids included
B851100WH White 10 x 75 cassettes (750)/cs Separate lids included
B851100YW Yellow 10 x 75 cassettes (750)/cs Separate lids included
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 48 options available.
48 options
Catalog No. B851010LA Supplier Epredia™ Supplier No. B851010LA
Only null left

Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.

Designed for performance and user convenience, PrintMate Embedding Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.PrintMate Embedding Cassettes are also available without lids, for economical use with reuseable metal lids.

Specifications

Pore Style Round Holes
Quantity 10 x 75 cassettes (750)/cs
Closure Included No lids
Closure Type Reversible
PrintMate Compatible Yes
Color Lavender
For Use With (Equipment) For Use with Metal Lids
Product Type Embedding Cassette