Learn More
Epredia™ PrintMate™ Biopsy Cassettes
Description
Designed for performance and user convenience, the PrintMate cassette range has been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low force opening and closing, while still giving the security needed while processing.Available in tubes for automatic loading or loose for manual use, and in twelve colors for enhanced workflow identification.
Specifications
Specifications
| Pore Style | Small Square Pores |
| Quantity | 4 x 250 cassettes (1000)/cs |
| Closure Included | Separate lids included |
| Closure Type | Reversible |
| PrintMate Compatible | Yes |
| Color | Beige |
| Product Type | PrintMate Biopsy Cassette for PrintMate Printers |