Learn More
Epredia™ PrintMate™ AS Cassette Printer
The small footprint of the Epredia™ SlideMateTM AS Slide Printer brings the convenience of on-demand slide printing directly to the microtome. The SlideMate AS Slide Printer drastically reduces the chance for patient identification errors by allowing the laboratory to print accurate information at the moment the slide is required.
Supplier:  Epredia™ A83800003
 View more versions of this product
Catalog No. A83800003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Prints each cassette in 10 seconds or less
Small footprint allows the printer to fit easily next to the grossing station
Print only the cassettes you need, right at the point of use Print barcodes on cassettes to automate downstream processes Eliminate manual handwriting processes
Specifications
Chemical Identifiers
100/240 VAC, 50/60 Hz, 150 VA (512 BTU/h)
Specifications
PrintMate AS 900 Cassette Printer
Cassette Printer
PrintMate AS
USB cable
Power cable
12 cassette hoppers
One roll of tape (installed)
Epredia™ LabWriter™ software CD
Operator guide
43.5 cm
38 cm
29 cm
14 kg with out Cassette Tubes 17 kg with Cassette Tubes
100/240 VAC, 50/60 Hz, 150 VA (512 BTU/h)
900 Cassettes
17 in.
15 in.
11.4 in.
31 lb. with out Cassette Tubes, 37.5 lb. with Cassette Tubes
100/240 VAC
Product Suggestions
Product Suggestions