Learn More
Epredia™ PrintMate™ AS Cassette Printer
The small footprint of the Epredia™ SlideMateTM AS Slide Printer brings the convenience of on-demand slide printing directly to the microtome. The SlideMate AS Slide Printer drastically reduces the chance for patient identification errors by allowing the laboratory to print accurate information at the moment the slide is required.
Supplier:  Epredia™ A83800002
 View more versions of this product
Catalog No. A83800002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Prints each cassette in 10 seconds or less
Small footprint allows the printer to fit easily next to the grossing station
Print only the cassettes you need, right at the point of use Print barcodes on cassettes to automate downstream processes Eliminate manual handwriting processes
Specifications
Chemical Identifiers
100/240 VAC, 50/60 Hz, 150 VA (512 BTU/h)
Specifications
PrintMate AS 450 Cassette Printer
Cassette Printer
PrintMate AS
USB cable
Power cable
6 cassette hoppers
One roll of tape (installed)
Epredia™ LabWriter™ software CD
Operator guide
100/240 VAC, 50/60 Hz, 150 VA (512 BTU/h)
450 Cassettes
Product Suggestions
Product Suggestions